Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320766.1 | NP_001307695.1 | 7 | 1117 | ||
NM_015565.2 | NP_056380.2 | 7 | 1114 | |||
XM_006723987.3 | XP_006724050.1 | 7 | 1112 | |||
XM_017028316.1 | XP_016883805.1 | 6 | 794 | |||
GenBank mRNA | AB018257.2 | - | 7 | 1005 | ||
AF161350.1 | - | 7 | 1001 | |||
AK001915.1 | - | 7 | 1061 | |||
AK023499.1 | - | 6 | 748 | |||
BC029371.1 | - | 7 | 1021 | |||
BC140790.1 | - | 7 | 1027 | 9021658 | ||
BC150284.1 | - | 7 | 1005 | 8860138 |