Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLITRK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015567.1 | NP_056382.1 | 2 | 3764 | ||
XM_005254038.4 | XP_005254095.1 | 2 | 4435 | |||
XM_005254039.2 | XP_005254096.1 | 2 | 4104 | |||
GenBank mRNA | AB020725.1 | - | 2 | 3570 | ||
AK024251.1 | - | 2 | 3765 |