Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127231.2 | NP_001120703.1 | 18 | 5124 | ||
NM_015570.3 | NP_056385.1 | 19 | 5196 | |||
XM_005250257.2 | XP_005250314.1 | 12 | 3270 | |||
XM_011516010.2 | XP_011514312.1 | 19 | 5661 | |||
XM_011516011.2 | XP_011514313.1 | 19 | 5658 | |||
XM_011516012.2 | XP_011514314.1 | 18 | 5595 | |||
XM_011516013.2 | XP_011514315.1 | 18 | 5589 | |||
XM_011516014.2 | XP_011514316.1 | 17 | 5559 | |||
XM_011516017.2 | XP_011514319.1 | 18 | 14215 | |||
XM_011516018.2 | XP_011514320.1 | 19 | 4196 | |||
GenBank mRNA | AB007902.2 | - | 20 | 4685 | ||
AF326917.1 | - | 19 | 4782 | |||
AK025298.1 | - | 1 | 1099 | |||
BC064693.1 | - | 18 | 5132 | 30345831 |