Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOC2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282062.1 | NP_001268991.1 | 10, 11 | 1198 | ||
NM_001282063.1 | NP_001268992.1 | 10, 11 | 1160 | |||
NM_001282068.1 | NP_001268997.1 | 9, 10 | 1125 | |||
NM_003586.2 | NP_003577.2 | 9, 10 | 1159 | |||
NR_104089.1 | 9, 10 | 942 | ||||
NR_104090.1 | 9, 10 | 879 | ||||
XM_011545975.1 | XP_011544277.1 | 11, 12 | 1584 | |||
XM_017023776.1 | XP_016879265.1 | 13, 14 | 1802 | |||
XM_017023777.1 | XP_016879266.1 | 10, 11 | 1739 | |||
XM_017023778.1 | XP_016879267.1 | 9, 10 | 1213 | |||
GenBank mRNA | AK293651.1 | - | 9, 10 | 828 | ||
AK309158.1 | - | 3, 4 | 477 | |||
BC041769.1 | - | 10, 11 | 1161 | 4819032 | ||
BC055284.1 | - | 9, 10 | 1159 | 5767656 | ||
BC063436.1 | - | 10, 11 | 1124 | 5394616 | ||
D31897.1 | - | 9, 10 | 1080 |