Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015626.8 | NP_056441.6 | 9 | 1440 | ||
NM_134265.2 | NP_599027.1 | 7 | 1002 | |||
XM_005257963.3 | XP_005258020.1 | 8 | 1237 | |||
XM_017024433.1 | XP_016879922.1 | 8 | 1326 | |||
XM_017024434.1 | XP_016879923.1 | 10 | 1528 | |||
XM_017024435.1 | XP_016879924.1 | 10 | 2895 | |||
XM_017024436.1 | XP_016879925.1 | 8 | 2627 | |||
GenBank mRNA | AF069313.2 | - | 9 | 1389 | ||
AF072880.1 | - | 9 | 1215 | |||
AF106683.1 | - | 9 | 1257 | |||
AF106684.1 | - | 4 | 989 | |||
AF112205.1 | - | 9 | 1379 | |||
AF240696.1 | - | 7 | 951 | |||
AK074917.1 | - | 9 | 1406 | |||
AK222576.1 | - | 9 | 1406 | |||
AL110243.1 | - | 9 | 1440 | |||
AL110269.1 | - | 9 | 2767 | |||
BC005226.1 | - | 4 | 442 | |||
BC021110.1 | - | 9 | 1387 | 4865448 | ||
BC048007.1 | - | 9 | 2762 |