Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015630.3 | NP_056445.3 | 3, 4 | 480 | ||
XM_011510941.2 | XP_011509243.1 | 3, 4 | 691 | |||
XM_011510943.2 | XP_011509245.1 | 2, 3 | 364 | |||
GenBank mRNA | AF286904.1 | - | 3, 4 | 472 | ||
AK001433.1 | - | 1, 2 | 71 | |||
AK125789.1 | - | 4, 5 | 549 | |||
BC093818.1 | - | 3, 4 | 392 | 7939663 | ||
BC093820.1 | - | 3, 4 | 392 | 7939665 |