Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015630.3 | NP_056445.3 | 6 | 933 | ||
XM_011510941.2 | XP_011509243.1 | 6 | 1144 | |||
XM_011510943.2 | XP_011509245.1 | 5 | 817 | |||
GenBank mRNA | AF286904.1 | - | 6 | 925 | ||
AK001433.1 | - | 4 | 524 | |||
AK125789.1 | - | 7 | 1002 | |||
BC093818.1 | - | 6 | 845 | 7939663 | ||
BC093820.1 | - | 6 | 845 | 7939665 |