Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164342.2 | NP_001157814.1 | 4 | 1610 | ||
NM_001164343.2 | NP_001157815.1 | 11 | 2072 | |||
NM_001164344.2 | NP_001157816.1 | 7 | 2036 | |||
NM_001164345.2 | NP_001157817.1 | 6 | 1990 | |||
NM_001164346.2 | NP_001157818.1 | 5 | 1629 | |||
NM_001164347.2 | NP_001157819.1 | 5 | 1651 | |||
NM_015642.5 | NP_056457.3 | 9 | 1834 | |||
GenBank mRNA | AF139460.2 | - | 4 | 1616 | ||
AK225788.1 | - | 6 | 1984 | |||
AK290116.1 | - | 2 | 1372 | |||
AK315307.1 | - | 5 | 1629 | |||
AL050276.1 | - | 7 | 1699 | |||
BC029041.1 | - | 5 | 1651 | 5184140 | ||
BC090936.1 | - | 11 | 2033 | |||
BX647778.1 | - | 9 | 1835 |