Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015666.3 | NP_056481.1 | 6 | 854 | ||
XM_005260393.1 | XP_005260450.1 | 6 | 844 | |||
XM_006723774.3 | XP_006723837.1 | 7 | 938 | |||
XM_011528776.1 | XP_011527078.1 | 6 | 2858 | |||
XM_017027805.1 | XP_016883294.1 | 5 | 3008 | |||
XM_017027806.1 | XP_016883295.1 | 6 | 843 | |||
XM_017027807.1 | XP_016883296.1 | 5 | 3008 | |||
XM_017027808.1 | XP_016883297.1 | 6 | 844 | |||
GenBank mRNA | AK001603.1 | - | 5 | 834 | ||
AK124572.1 | - | 6 | 3145 | |||
AK299147.1 | - | 4 | 512 | |||
AK299189.1 | - | 5 | 721 | |||
AK316027.1 | - | 5 | 634 | |||
AK316325.1 | - | 6 | 706 | |||
BC012073.1 | - | 4 | 719 | |||
BC036716.2 | - | 5 | 828 | 4900170 |