Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015666.3 | NP_056481.1 | 6, 7 | 859 | ||
XM_005260393.1 | XP_005260450.1 | 6, 7 | 849 | |||
XM_006723774.3 | XP_006723837.1 | 7, 8 | 943 | |||
XM_011528776.1 | XP_011527078.1 | 6, 7 | 2863 | |||
XM_017027805.1 | XP_016883294.1 | 5, 6 | 3013 | |||
XM_017027806.1 | XP_016883295.1 | 6, 7 | 848 | |||
GenBank mRNA | AK001603.1 | - | 5, 6 | 839 | ||
AK124572.1 | - | 6, 7 | 3150 | |||
AK299147.1 | - | 4, 5 | 517 | |||
AK299189.1 | - | 5, 6 | 726 | |||
AK316027.1 | - | 5, 6 | 639 | |||
AK316325.1 | - | 6, 7 | 711 | |||
BC012073.1 | - | 4, 5 | 724 | |||
BC036716.2 | - | 5, 6 | 833 | 4900170 |