Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321389.1 | NP_001308318.1 | 9 | 1826 | ||
NM_001321390.1 | NP_001308319.1 | 9 | 1471 | |||
NM_001321391.1 | NP_001308320.1 | 9 | 1505 | |||
NM_015680.5 | NP_056495.3 | 8 | 1563 | |||
NR_135628.1 | 8 | 1443 | ||||
NR_135629.1 | 7 | 1501 | ||||
GenBank mRNA | AF070638.1 | - | 2 | 825 | ||
AF151815.1 | - | 9 | 1730 | |||
AK075274.1 | - | 8 | 1519 | |||
AY007160.1 | - | 8 | 1536 | |||
BC001393.2 | - | 8 | 1486 | 3051397 | ||
BC012821.2 | - | 8 | 1535 | 2958621 |