Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199784.1 | NP_001186713.1 | 9 | 1302 | ||
NM_015685.5 | NP_056500.2 | 5 | 1193 | |||
NM_080489.4 | NP_536737.3 | 9 | 1413 | |||
GenBank mRNA | AF131809.1 | - | 5 | 1159 | ||
AF159228.1 | - | 9 | 1302 | |||
BC002727.2 | - | 9 | 1352 | 3633262 |
Gene Symbol: FKBP1A-SDCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037661.1 | 10 | 1521 |