Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324403.1 | NP_001311332.1 | 2 | 351 | ||
NM_015886.4 | NP_056970.1 | 2 | 323 | |||
GenBank mRNA | BC074931.2 | - | 1 | 167 | 30915489 | |
BC074932.2 | - | 1 | 167 | 30915266 | ||
BC126290.1 | - | 1 | 172 | 8992006 | ||
BC126292.1 | - | 1 | 174 | 8992008 | ||
CR749657.1 | - | 2 | 324 | |||
D45027.1 | - | 1 | 628 |