Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MED15 SIRNA Assays ›
Gene Symbol:
MED15
Gene Name:
mediator complex subunit 15
Gene Aliases:
ARC105, CAG7A, CTG7A, PCQAP, TIG-1, TIG1, TNRC7
Chromosome Location:
Chr.22: 20507542 - 20587632 on Build GRCh38
UniGene ID:
Hs.517421
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
134690
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MED15
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003891.2
NP_001003891.1
11
1661
NM_001293234.1
NP_001280163.1
10
1490
NM_001293235.1
NP_001280164.1
10
1530
NM_001293236.1
NP_001280165.1
9
1328
NM_001293237.1
NP_001280166.1
10
1733
NM_015889.4
NP_056973.2
10
1541
XM_006724263.3
XP_006724326.1
10
1406
XM_006724264.3
XP_006724327.1
11
1632
XM_011530214.2
XP_011528516.1
12
1705
XM_011530215.2
XP_011528517.1
10
1527
XM_017028817.1
XP_016884306.1
10
1603
XM_017028818.1
XP_016884307.1
12
1803
XM_017028819.1
XP_016884308.1
11
1569
XM_017028820.1
XP_016884309.1
10
1512
XM_017028823.1
XP_016884312.1
10
2784
XM_017028824.1
XP_016884313.1
12
1701
GenBank mRNA
AF056191.1
-
10
1490
AF328769.1
-
10
1398
AK000003.1
-
1
861
AK074230.1
-
5
688
AK074268.1
-
10
1335
AK090465.1
-
11
1593
AK124276.1
-
3
1807
AK124925.1
-
3
1012
AK126208.1
-
6
1866
AK294540.1
-
12
1537
AK294734.1
-
10
1729
AK315540.1
-
10
1453
AL832151.1
-
3
676
BC003078.1
-
10
1462
BC007529.1
-
3
314
BC013985.2
-
10
1444
3506327
BC017110.1
-
7
1289
CR456537.1
-
9
1220
U80745.1
-
5
852
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
RNA polymerase II transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0