Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015891.2 | NP_056975.1 | 3 | 381 | ||
GenBank mRNA | AB208903.1 | - | 3 | 355 | ||
AF015044.1 | - | 3 | 348 | |||
AF038392.1 | - | 3 | 381 | |||
AF061241.1 | - | 3 | 354 | |||
AK222721.1 | - | 3 | 378 | |||
AK295055.1 | - | 3 | 374 | |||
AK314601.1 | - | 3 | 377 | |||
BC111061.1 | - | 4 | 458 | |||
BC117145.1 | - | 3 | 378 | 40125696 | ||
BC126114.1 | - | 3 | 378 | 8991830 |