Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015896.3 | NP_056980.2 | 7 | 789 | ||
XM_005265216.2 | XP_005265273.1 | 6 | 962 | |||
GenBank mRNA | AB209621.1 | - | 1 | 3531 | ||
AK058135.1 | - | 7 | 719 | |||
AK093455.1 | - | 1 | 173 | |||
AK096525.1 | - | 7 | 1897 | |||
AK223343.1 | - | 7 | 704 | |||
AK308086.1 | - | 7 | 686 | |||
AL833828.1 | - | 7 | 718 | |||
BC033732.1 | - | 7 | 685 | 5168611 | ||
U70824.1 | - | 7 | 743 |