Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015906.3 | NP_056990.3 | 7 | 1366 | ||
NM_033020.2 | NP_148980.2 | 7 | 1366 | |||
XM_005270936.3 | XP_005270993.1 | 7 | 1287 | |||
XM_005270937.3 | XP_005270994.1 | 7 | 1287 | |||
XM_011541568.2 | XP_011539870.1 | 7 | 1287 | |||
XM_017001452.1 | XP_016856941.1 | 5 | 641 | |||
XM_017001453.1 | XP_016856942.1 | 5 | 641 | |||
XM_017001454.1 | XP_016856943.1 | 7 | 1287 | |||
GenBank mRNA | AB029036.1 | - | 7 | 1295 | ||
AF119043.1 | - | 7 | 1366 | |||
AF220136.1 | - | 7 | 1366 | |||
AF220137.1 | - | 7 | 1366 | |||
AJ132948.1 | - | 7 | 1057 | |||
AK023434.1 | - | 4 | 507 | |||
AK301997.1 | - | 1 | 26 |