Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015963.5 | NP_057047.4 | 2 | 1321 | ||
XM_005247016.4 | XP_005247073.3 | 2 | 1554 | |||
XM_011511291.2 | XP_011509593.1 | 2 | 1123 | |||
XM_017004256.1 | XP_016859745.1 | 2 | 1115 | |||
GenBank mRNA | AF258556.1 | - | 2 | 1073 | ||
AK001216.1 | - | 1 | 368 | |||
BC000247.1 | - | 1 | 763 | |||
BC000767.2 | - | 1 | 236 | |||
BC001842.2 | - | 1 | 236 | |||
BC009439.1 | - | 1 | 859 | |||
BC069235.1 | - | 2 | 1251 | 6645198 | ||
BC071896.1 | - | 1 | 287 | |||
BC094822.1 | - | 1 | 287 |