Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015971.3 | NP_057055.2 | 4 | 637 | ||
GenBank mRNA | AF077042.1 | - | 4 | 586 | ||
AK222995.1 | - | 4 | 601 | |||
AK313854.1 | - | 4 | 411 | |||
BC000241.1 | - | 4 | 596 | 3350465 | ||
BC050641.2 | - | 4 | 393 | |||
BU599633.1 | - | 4 | 615 |