Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015975.4 | NP_057059.2 | 7 | 884 | ||
GenBank mRNA | AF220509.1 | - | 7 | 835 | ||
BC009566.1 | - | 7 | 835 | 3996687 | ||
BC010350.1 | - | 7 | 849 | 4276335 | ||
BC071649.1 | - | 7 | 821 | 5246715 | ||
BC146952.1 | - | 7 | 865 | 9056880 | ||
BC146960.1 | - | 7 | 865 | 9056888 |