Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015976.4 | NP_057060.2 | 5 | 784 | ||
NM_152238.3 | NP_689424.2 | 4 | 619 | |||
NR_033716.1 | 6 | 904 | ||||
XM_006710677.2 | XP_006710740.1 | 5 | 801 | |||
XM_006710678.2 | XP_006710741.1 | 6 | 900 | |||
XM_011541564.2 | XP_011539866.1 | 6 | 840 | |||
XM_017001425.1 | XP_016856914.1 | 5 | 762 | |||
XM_017001426.1 | XP_016856915.1 | 6 | 899 | |||
XM_017001427.1 | XP_016856916.1 | 6 | 1174 | |||
XM_017001428.1 | XP_016856917.1 | 5 | 649 | |||
GenBank mRNA | AF121857.1 | - | 5 | 627 | ||
AK025031.1 | - | 5 | 741 | |||
AK223229.1 | - | 5 | 643 | |||
AK293018.1 | - | 5 | 712 | |||
AK298211.1 | - | 4 | 465 | |||
AL049989.1 | - | 5 | 641 | |||
AL548028.3 | - | 5 | 785 | |||
BC010349.1 | - | 5 | 598 | 4093672 | ||
BC018105.1 | - | 6 | 722 | 3849824 | ||
BT006747.1 | - | 4 | 540 |