Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015993.2 | NP_057077.1 | 4 | 1077 | ||
GenBank mRNA | AF137386.1 | - | 4 | 1073 | ||
AK130584.1 | - | 4 | 1069 | |||
AK289599.1 | - | 4 | 1054 | |||
BC002760.2 | - | 4 | 1040 | 3633345 |