Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008222.2 | NP_001008223.1 | 4 | 837 | ||
NM_016032.3 | NP_057116.2 | 5 | 817 | |||
XM_011531348.2 | XP_011529650.1 | 5 | 804 | |||
GenBank mRNA | AB209807.1 | - | 4 | 756 | ||
AF151847.1 | - | 5 | 705 | |||
AK001112.1 | - | 5 | 767 | |||
AK001424.1 | - | 2 | 151 | |||
AK001524.1 | - | 4 | 568 | |||
AK096967.1 | - | 5 | 468 | |||
AL161962.1 | - | 5 | 694 | |||
AY358558.1 | - | 5 | 749 | |||
AY952881.8 | - | 5 | 789 | |||
BC000035.2 | - | 4 | 711 | 3506601 | ||
BC003128.1 | - | 5 | 755 | 2957911 | ||
BC006200.2 | - | 5 | 751 | 2964425 | ||
BC012826.2 | - | 5 | 751 | 2964425 |