Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270879.1 | NP_001257808.1 | 6 | 1063 | ||
NM_016098.3 | NP_057182.1 | 5 | 847 | |||
NR_073093.1 | 6 | 1076 | ||||
XM_011535894.1 | XP_011534196.1 | 5 | 1846 | |||
XM_011535895.1 | XP_011534197.1 | 4 | 1630 | |||
GenBank mRNA | AF125101.1 | - | 5 | 832 | ||
AF151887.1 | - | 5 | 829 | |||
AF275811.1 | - | 3 | 554 | |||
AL556427.3 | - | 5 | 766 | |||
BC000810.1 | - | 5 | 757 | 3452973 |