Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016107.3 | NP_057191.2 | 2 | 162 | ||
GenBank mRNA | AJ314790.1 | - | 2 | 115 | ||
AK292214.1 | - | 2 | 162 | |||
AK309638.1 | - | 2 | 162 | |||
BC017879.1 | - | 2 | 162 | |||
BC030540.1 | - | 2 | 134 | |||
BC107417.1 | - | 2 | 113 | |||
BC137084.1 | - | 2 | 126 | 9021081 | ||
BC137085.1 | - | 2 | 126 | 9021082 |