Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016107.3 | NP_057191.2 | 13 | 2260 | ||
GenBank mRNA | AF100742.1 | - | 3 | 255 | ||
AJ314790.1 | - | 13 | 2213 | |||
BC051893.1 | - | 6 | 851 | |||
BC062986.1 | - | 6 | 776 | |||
BC137084.1 | - | 13 | 2224 | 9021081 | ||
BC137085.1 | - | 13 | 2224 | 9021082 |