Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201965.1 | NP_001188894.1 | 7 | 1493 | ||
NM_016115.4 | NP_057199.1 | 8 | 1327 | |||
NM_145863.2 | NP_665862.1 | 7 | 1118 | |||
GenBank mRNA | AB209795.1 | - | 8 | 1226 | ||
AF156778.1 | - | 7 | 1125 | |||
AK000985.1 | - | 8 | 1253 | |||
AK001283.1 | - | 7 | 1051 | |||
AK056069.1 | - | 7 | 1495 | |||
AK222595.1 | - | 7 | 1137 | |||
AK222715.1 | - | 7 | 1109 | |||
AK303133.1 | - | 6 | 1010 | |||
BC006488.2 | - | 7 | 1139 | 3537359 | ||
BC009569.1 | - | 8 | 1185 | 4052577 |
Gene Symbol: GPR75-ASB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164165.1 | NP_001157637.1 | 8 | 1344 | ||
GenBank mRNA | BC110915.1 | - | 8 | 1266 | 5764788 |