Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016129.2 | NP_057213.2 | 9 | 1183 | ||
XM_011532019.1 | XP_011530321.1 | 9 | 1179 | |||
GenBank mRNA | AF100757.1 | - | 9 | 1047 | ||
AK001148.1 | - | 9 | 1179 | |||
AK001210.1 | - | 9 | 1010 | |||
AK024005.1 | - | 9 | 1068 | |||
AK223178.1 | - | 9 | 1053 | |||
BC004302.1 | - | 9 | 1046 | 3623514 | ||
BC009292.2 | - | 9 | 1046 | 4111685 | ||
BC093007.1 | - | 9 | 1068 | 30340740 | ||
KU178633.1 | - | 9 | 1044 | |||
KU178634.1 | - | 9 | 1044 |