Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSIG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321329.1 | NP_001308258.1 | 6 | 2104 | ||
NM_001321330.1 | NP_001308259.1 | 5 | 1729 | |||
NM_001321331.1 | NP_001308260.1 | 6 | 1737 | |||
NM_001321332.1 | NP_001308261.1 | 6 | 1730 | |||
NM_001321333.1 | NP_001308262.1 | 5 | 1637 | |||
NM_016133.3 | NP_057217.2 | 6 | 2111 | |||
GenBank mRNA | AF527632.1 | - | 6 | 2060 | ||
AK021692.1 | - | 5 | 1543 | |||
AL080184.1 | - | 2 | 1246 | |||
BC022475.1 | - | 6 | 2055 | 4794938 | ||
BX647805.1 | - | 6 | 1653 |