Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016148.3 | NP_057232.2 | 22 | 4895 | ||
XM_006723233.3 | XP_006723296.1 | 23 | 5293 | |||
XM_011527013.2 | XP_011525315.1 | 24 | 5317 | |||
XM_011527014.2 | XP_011525316.1 | 22 | 5266 | |||
GenBank mRNA | AF163302.1 | - | 22 | 4895 | ||
AF226728.1 | - | 8 | 3037 |