Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUFU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178133.1 | NP_001171604.1 | 9 | 1294 | ||
NM_016169.3 | NP_057253.2 | 9 | 1294 | |||
XM_011539858.2 | XP_011538160.1 | 9 | 1297 | |||
XM_011539860.2 | XP_011538162.1 | 9 | 1294 | |||
XM_011539861.2 | XP_011538163.1 | 9 | 1297 | |||
XM_011539863.2 | XP_011538165.1 | 9 | 1032 | |||
XM_011539864.2 | XP_011538166.1 | 9 | 1297 | |||
XM_017016323.1 | XP_016871812.1 | 9 | 1527 | |||
GenBank mRNA | AF144231.1 | - | 9 | 1249 | ||
AF159447.1 | - | 9 | 1249 | |||
AF172319.1 | - | 9 | 1099 | |||
AF175770.1 | - | 9 | 1269 | |||
AF222345.1 | - | 9 | 1177 | |||
AK307370.1 | - | 4 | 503 | |||
AK311503.1 | - | 4 | 472 | |||
AK312319.1 | - | 9 | 1284 | |||
AY358550.1 | - | 9 | 1249 | |||
BC013291.2 | - | 9 | 1270 | 3533158 |