Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRTO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016183.3 | NP_057267.2 | 3 | 370 | ||
XM_006710675.3 | XP_006710738.1 | 3 | 528 | |||
GenBank mRNA | AF173378.1 | - | 3 | 145 | ||
AK024227.1 | - | 3 | 131 | |||
AK027569.1 | - | 3 | 146 | |||
AK295328.1 | - | 3 | 117 | |||
AY303790.1 | - | 3 | 126 | |||
BC003013.1 | - | 3 | 144 | 2821148 | ||
BC006504.2 | - | 3 | 144 | 2821148 | ||
BQ670859.1 | - | 3 | 325 |