Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016184.3 | NP_057268.1 | 6 | 991 | ||
NM_194447.2 | NP_919429.2 | 5 | 874 | |||
NM_194448.2 | NP_919430.1 | 4 | 775 | |||
NM_194450.2 | NP_919432.1 | 5 | 892 | |||
XM_011520684.2 | XP_011518986.1 | 5 | 2252 | |||
XM_017019382.1 | XP_016874871.1 | 7 | 912 | |||
GenBank mRNA | AA418441.1 | - | 2 | 226 | ||
AF067800.1 | - | 6 | 1012 | |||
AF109146.1 | - | 6 | 838 | |||
AF328684.1 | - | 6 | 984 | |||
AJ133532.1 | - | 6 | 985 | |||
BC117439.1 | - | 6 | 886 | 40125990 | ||
BC117441.1 | - | 6 | 886 | 40125992 |