Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMOTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278683.1 | NP_001265612.1 | 10 | 4593 | ||
NM_001278685.1 | NP_001265614.1 | 10 | 4601 | |||
NM_016201.3 | NP_057285.3 | 10 | 4610 | |||
GenBank mRNA | AW294642.1 | - | 1 | 317 |