Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003654.5 | NP_003645.1 | 4 | 1983 | ||
GenBank mRNA | AB003791.1 | - | 4 | 1644 | ||
BC022567.1 | - | 4 | 1949 | 4814499 | ||
BC028235.1 | - | 4 | 1617 | 5245338 | ||
U65637.1 | - | 2 | 1402 |