Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016224.4 | NP_057308.1 | 6 | 809 | ||
XM_005267015.2 | XP_005267072.1 | 8 | 887 | |||
XM_011535886.2 | XP_011534188.1 | 3 | 348 | |||
GenBank mRNA | AB209736.1 | - | 2 | 140 | ||
AF001629.1 | - | 6 | 614 | |||
AF121859.1 | - | 6 | 569 | |||
AF131214.1 | - | 6 | 588 | |||
AF172847.1 | - | 6 | 689 | |||
AK026637.1 | - | 3 | 354 | |||
AK127345.1 | - | 3 | 664 | |||
AK314360.1 | - | 6 | 643 | |||
BC005022.1 | - | 6 | 716 | 3832234 |