Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016224.4 | NP_057308.1 | 6 | 846 | ||
XM_005267015.2 | XP_005267072.1 | 8 | 924 | |||
XM_011535886.2 | XP_011534188.1 | 3 | 385 | |||
GenBank mRNA | AB209736.1 | - | 2 | 177 | ||
AF001629.1 | - | 6 | 651 | |||
AF076957.1 | - | 1 | 66 | |||
AF121859.1 | - | 6 | 606 | |||
AF131214.1 | - | 6 | 625 | |||
AF172847.1 | - | 6 | 726 | |||
AK026637.1 | - | 3 | 391 | |||
AK127345.1 | - | 3 | 701 | |||
AK314360.1 | - | 6 | 680 | |||
BC005022.1 | - | 6 | 753 | 3832234 |