Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016231.4 | NP_057315.3 | 1 | 625 | ||
XM_005257988.2 | XP_005258045.1 | 1 | 640 | |||
GenBank mRNA | AF180819.1 | - | 1 | 623 | ||
AF197898.1 | - | 1 | 625 | |||
AK024686.1 | - | 1 | 257 | |||
AK315315.1 | - | 1 | 640 | |||
BC064663.1 | - | 1 | 1137 | 6527673 | ||
DA224552.1 | - | 1 | 486 |