Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137559.1 | NP_001131031.1 | 4 | 354 | ||
NM_016237.4 | NP_057321.2 | 4 | 576 | |||
XM_005253900.3 | XP_005253957.1 | 4 | 578 | |||
XM_017019423.1 | XP_016874912.1 | 4 | 294 | |||
XM_017019424.1 | XP_016874913.1 | 4 | 294 | |||
GenBank mRNA | AF191339.1 | - | 4 | 454 | ||
AK025614.1 | - | 4 | 354 | |||
AK315451.1 | - | 4 | 522 | |||
BC001081.2 | - | 4 | 509 | 2823401 | ||
BC001950.1 | - | 4 | 509 | 2823401 | ||
BC006301.2 | - | 4 | 513 | 4100632 | ||
BC034243.1 | - | 3 | 216 | |||
BX537687.1 | - | 1 | 257 |