Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016245.4 | NP_057329.3 | 7 | 1136 | ||
GenBank mRNA | AF126780.1 | - | 7 | 1091 | ||
AF273056.1 | - | 7 | 989 | |||
AK075348.1 | - | 7 | 1020 | |||
AY358553.1 | - | 7 | 1003 | |||
BC008650.1 | - | 7 | 1026 | 3854750 | ||
BC014327.1 | - | 7 | 1002 | 3961721 | ||
BC016367.1 | - | 7 | 1033 | 4133318 | ||
BC021673.1 | - | 7 | 1007 | 3827935 | ||
BC036001.1 | - | 7 | 1027 | 4715042 |