Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSL24D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016304.2 | NP_057388.1 | 2 | 323 | ||
GenBank mRNA | AB462502.1 | - | 2 | 207 | ||
AF060926.1 | - | 2 | 190 | |||
AF165521.1 | - | 2 | 323 | |||
AF201949.1 | - | 2 | 323 | |||
AF212226.1 | - | 2 | 323 | |||
AK055713.1 | - | 2 | 210 | |||
AK315431.1 | - | 2 | 223 | |||
BC008409.1 | - | 2 | 245 | 4097130 | ||
BC008422.1 | - | 2 | 215 | 4096889 | ||
BC008449.1 | - | 2 | 221 | 4133589 | ||
BC008499.1 | - | 2 | 236 | 4286115 | ||
BC009593.1 | - | 2 | 226 | 4109097 | ||
BC009604.1 | - | 2 | 226 | 4282059 | ||
BC012913.1 | - | 2 | 190 | 4514979 | ||
BC014576.1 | - | 2 | 225 | 3930303 | ||
BC016312.1 | - | 2 | 231 | 4096867 | ||
BC016331.1 | - | 2 | 231 | 4108672 | ||
BC016725.1 | - | 2 | 232 | 4078305 | ||
BC016777.1 | - | 2 | 213 | 4104810 | ||
BC026266.1 | - | 2 | 227 | 4276092 | ||
BC026267.1 | - | 2 | 206 | 4277062 | ||
BC028672.1 | - | 2 | 212 | 4092804 | ||
BC035995.1 | - | 2 | 236 | 4671856 | ||
BC093067.1 | - | 2 | 210 | 30419021 | ||
CR457179.1 | - | 2 | 180 |