Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSL24D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016304.2 | NP_057388.1 | 6 | 1151 | ||
GenBank mRNA | AF060926.1 | - | 6 | 1014 | ||
AF212226.1 | - | 6 | 1151 | |||
AK055713.1 | - | 6 | 1037 | |||
BC008409.1 | - | 6 | 1074 | 4097130 | ||
BC008422.1 | - | 6 | 1041 | 4096889 | ||
BC008449.1 | - | 6 | 1049 | 4133589 | ||
BC009593.1 | - | 6 | 1054 | 4109097 | ||
BC009604.1 | - | 6 | 1055 | 4282059 | ||
BC012913.1 | - | 6 | 1018 | 4514979 | ||
BC016312.1 | - | 6 | 1059 | 4096867 | ||
BC016331.1 | - | 6 | 1059 | 4108672 | ||
BC016725.1 | - | 6 | 1056 | 4078305 | ||
BC016777.1 | - | 6 | 1041 | 4104810 | ||
BC026266.1 | - | 6 | 1055 | 4276092 | ||
BC026267.1 | - | 6 | 1034 | 4277062 | ||
BC028672.1 | - | 6 | 1040 | 4092804 | ||
BC035995.1 | - | 6 | 1064 | 4671856 | ||
BC093067.1 | - | 6 | 1033 | 30419021 |