Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003673.3 | NP_003664.1 | 1 | 100 | ||
GenBank mRNA | AJ000491.1 | - | 1 | 96 | ||
AK096328.1 | - | 1 | 1259 | |||
BC012628.1 | - | 1 | 101 | 4251080 | ||
BC013330.1 | - | 1 | 101 | 4273711 | ||
BF670957.1 | - | 1 | 101 | |||
DA899335.1 | - | 1 | 100 | |||
KU178429.1 | - | 1 | 86 | |||
KU178430.1 | - | 1 | 86 |