Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016337.2 | NP_057421.1 | 6 | 794 | ||
XM_005267749.3 | XP_005267806.1 | 6 | 1413 | |||
XM_011536828.2 | XP_011535130.1 | 6 | 1427 | |||
XM_017021363.1 | XP_016876852.1 | 6 | 1413 | |||
XM_017021364.1 | XP_016876853.1 | 6 | 1300 | |||
GenBank mRNA | AF052504.1 | - | 6 | 768 | ||
AF087843.1 | - | 6 | 773 | |||
AF112209.1 | - | 6 | 887 | |||
AF131766.1 | - | 4 | 541 | |||
AK289720.1 | - | 7 | 804 | |||
AK295919.1 | - | 6 | 1002 | |||
AK301379.1 | - | 7 | 675 | |||
AL133642.1 | - | 1 | 216 | |||
BC023997.2 | - | 6 | 943 | 2822488 | ||
BC032358.1 | - | 6 | 765 | 5222624 |