Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016337.2 | NP_057421.1 | 14 | 1644 | ||
XM_005267749.3 | XP_005267806.1 | 14 | 2263 | |||
XM_017021364.1 | XP_016876853.1 | 14 | 2150 | |||
GenBank mRNA | AF052504.1 | - | 14 | 1618 | ||
AF087843.1 | - | 14 | 1623 | |||
AF112209.1 | - | 14 | 1737 | |||
AF131766.1 | - | 12 | 1391 | |||
AK091634.1 | - | 1 | 2120 | |||
AK092467.1 | - | 3 | 1349 | |||
BC023997.2 | - | 14 | 1793 | 2822488 | ||
BC032358.1 | - | 14 | 1615 | 5222624 | ||
BC049376.1 | - | 3 | 1278 |