Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPO11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134779.1 | NP_001128251.1 | 7, 8 | 960 | ||
NM_016338.4 | NP_057422.3 | 7, 8 | 900 | |||
GenBank mRNA | AF111109.1 | - | 6, 7 | 710 | ||
AK001696.1 | - | 7, 8 | 882 | |||
BC033776.1 | - | 7, 8 | 862 | 4525238 |