Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016353.4 | NP_057437.1 | 13 | 2802 | ||
XM_011544544.2 | XP_011542846.1 | 15 | 3232 | |||
XM_011544545.2 | XP_011542847.1 | 14 | 3145 | |||
XM_011544546.2 | XP_011542848.1 | 15 | 2423 | |||
XM_011544547.2 | XP_011542849.1 | 14 | 2506 | |||
XM_011544548.2 | XP_011542850.1 | 12 | 2268 | |||
GenBank mRNA | AB023584.1 | - | 13 | 2802 | ||
AK001608.1 | - | 6 | 1737 | |||
BC039253.1 | - | 13 | 2608 | 5299722 |