Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002026.2 | NP_001002026.1 | 5 | 835 | ||
NM_016369.3 | NP_057453.1 | 5 | 844 | |||
GenBank mRNA | AF221069.1 | - | 5 | 791 | ||
AK098474.1 | - | 5 | 835 | |||
AK298103.1 | - | 6 | 867 | |||
AY358479.1 | - | 5 | 871 | |||
BC142708.1 | - | 5 | 850 | 40148818 | ||
BC146668.1 | - | 5 | 850 | 40148169 |