Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257999.1 | NP_001244928.1 | 5 | 719 | ||
NM_016376.3 | NP_057460.3 | 5 | 514 | |||
NR_047571.1 | 4 | 395 | ||||
XM_005256679.3 | XP_005256736.1 | 5 | 561 | |||
XM_011523926.1 | XP_011522228.1 | 5 | 617 | |||
XM_011523928.2 | XP_011522230.1 | 5 | 599 | |||
XM_017024733.1 | XP_016880222.1 | 5 | 814 | |||
GenBank mRNA | AB033081.2 | - | 5 | 719 | ||
AB037360.1 | - | 5 | 482 | |||
AK025483.1 | - | 5 | 491 | |||
AK057047.1 | - | 4 | 372 | |||
AK225170.1 | - | 5 | 491 | |||
AK292930.1 | - | 5 | 491 | |||
AL706226.1 | - | 5 | 514 | |||
BC052308.1 | - | 5 | 487 | |||
DA856905.1 | - | 5 | 514 |