Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198976.2 | NP_945327.2 | 10 | 1318 | ||
GenBank mRNA | AF161479.1 | - | 10 | 1250 | ||
AJ238374.1 | - | 5 | 715 | |||
AJ238375.1 | - | 5 | 641 | |||
AJ238379.1 | - | 10 | 1252 | |||
AK001316.1 | - | 10 | 1226 | |||
AK023927.1 | - | 10 | 1243 | |||
AK127431.1 | - | 1 | 1914 | |||
AK293410.1 | - | 10 | 1270 | |||
BC014952.1 | - | 10 | 1254 | 4860894 | ||
BX647417.1 | - | 10 | 1253 |